missing translation for 'onlineSavingsMsg'
Learn More
Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals Product Code.: 11337477

Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals

Product Code. 11337477
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11337477

Brand: Thermo Scientific Alfa Aesar 012628.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Oxidation

Ruthenium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314075-07-5
Molecular Formula Br3H2ORu
Molecular Weight (g/mol) 358.80
MDL Number MFCD00016314
InChI Key PMMMCGISKBNZES-UHFFFAOYSA-K
IUPAC Name tribromoruthenium hydrate
SMILES O.Br[Ru](Br)Br

Specifications

Physical Form Crystalline
Quantity 1 g
UN Number UN3260
Solubility Information Soluble in alcohol
Formula Weight 340.78 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ru 25% min.
Odor Characteristic
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) bromide hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX26H016 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012628.03,012628.06
Lot NumberP23I053 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012628.03,012628.06
Lot NumberP08I011 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012628.03,012628.06
Lot NumberC20Z024 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012628.03,012628.06
Lot NumberK22M33 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012628.03,012628.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.