missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) bromide hydrate, Ru 25% min, Thermo Scientific Chemicals

Product Code. 11347477
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Oxidation

Ruthenium(III) bromide hydrate is used as a catalyst in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314075-07-5
MDL Number MFCD00016314

Specifications

Physical Form Crystalline
Quantity 5 g
UN Number UN3260
Solubility Information Soluble in alcohol
Formula Weight 340.78 (Anhydrous)
Concentration or Composition (by Analyte or Components) Ru 25% min.
Odor Characteristic
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) bromide hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'