missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) bromide, Ru 29% min, Thermo Scientific Chemicals

Product Code. 11351848
Click to view available options
Quantity:
5 g

Ruthenium(III) bromide is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14014-88-1
MDL Number MFCD00016314

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3260
Solubility Information Insoluble in water.
Formula Weight 340.78
Concentration or Composition (by Analyte or Components) Ru 29% min.
Odor Characteristic
Sensitivity Hygroscopic
Density 5.30 g/mL
Chemical Name or Material Ruthenium(III) bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'