missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis

Specifications
Specifications
Melting Point | >500.0°C |
Color | Black |
Physical Form | Crystals or Powder |
Quantity | 2 g |
Assay Percent Range | 48 to 50% (Ru) |
Linear Formula | RuCl3 |
Fieser | 04,421; 08,437; 10,343; 13,268; 14,271; 15,280; 16,292 |
Merck Index | 15,8436 |
Solubility Information | Solubility in water: insoluble. Other solubilities: decomposes in hot water, slightly soluble in alcohol |
Formula Weight | 207.43 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464261 | Certificate of Analysis | 16/07/2024 | 197210100,197210020,197210500 |
A0462555 | Certificate of Analysis | 07/05/2024 | 197210100,197210020,197210500 |
A0458807 | Certificate of Analysis | 15/12/2023 | 197210100,197210020,197210500 |
A0442229 | Certificate of Analysis | 25/08/2023 | 197210100,197210020,197210500 |
A0443923 | Certificate of Analysis | 25/08/2023 | 197210100,197210020,197210500 |
Safety and Handling



Product Identifier
- Ruthenium(III) chloride
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- ACUTE AQUATIC Acute 1
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H400-Very toxic to aquatic life.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ruthenium(III) chloride, 99.9%, (trace metal basis), anhydrous, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction