Learn More
Ruthenium(III) chloride hydrate, 99.9% (PGM basis), Ru 38% min, Thermo Scientific Chemicals
Description
Oxidation.Ruthenium(III) chloride hydrate is used as a catalyst for alkyne functionalization and oxidation reactions. It is used as key raw materials for the preparation of various ruthenium-based catalysts, including Grubbs’ catalysts. It is utilized in the preparation of nanoparticulate ruthenium-aluminum oxyhydroxide catalyst, which is efficient for dehydrogenation of alcohols to the corresponding aldehydes. It is also involved in the synthesis of 2-ethyl-3-methylquinolines from primary aromatic amines and triallylamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100°dec |
Physical Form | Crystalline Soluble |
Quantity | 2 g |
Assay Percent Range | 99.9% |
Solubility Information | Very soluble in water. Soluble in alcohol,acetone. |
Formula Weight | 207.43 (Anhydrous) |
Odor | Pungent |
Sensitivity | Hygroscopic |
Appearance | Black crystalline or powder |
Chemical Name or Material | Ruthenium(III) chloride hydrate, PGM basis, Ru 38% min. |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X11K024 | Certificate of Analysis | 30/10/2024 | 011043.09,011043.18,011043.04 |
M25K024 | Certificate of Analysis | 01/02/2024 | 011043.09,011043.18,011043.04 |
M04K025 | Certificate of Analysis | 22/01/2024 | 011043.09,011043.18,011043.04 |
T24J072 | Certificate of Analysis | 14/08/2023 | 011043.09,011043.18,011043.04 |
Q21J010 | Certificate of Analysis | 01/05/2023 | 011043.09,011043.18,011043.04 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.