Learn More
Ruthenium(III) chloride oxide, ammoniated, Thermo Scientific Chemicals
Microscopic stain
85.80 € - 353.00 €
Identifiants chimiques
CAS | 11103-72-3 |
---|---|
Molecular Formula | Cl6H46N14O2Ru3 |
Molecular Weight (g/mol) | 790.374 |
MDL Number | MFCD00011479 |
InChI Key | JQJSTVUROJELSR-UHFFFAOYSA-H |
Synonym | 6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride |
PubChem CID | 16218584 |
IUPAC Name | azane;ruthenium(2+);hexachloride;dihydrate |
SMILES | N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] |
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
11341848
|
Thermo Scientific Alfa Aesar
011806.03 |
1 g |
85.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331848
|
Thermo Scientific Alfa Aesar
011806.06 |
5 g |
353.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Microscopic stainRuthenium(III) chloride oxide, ammoniated is used as a reagent on plant material for staining pectin, gum, animal tissues and bacteria. It is also used in histology to stain aldehyde fixed mucopolysaccharides. Further, it serves as a pharmacological tool to study specific cellular mechanisms. It serves as an inhibitor of mitochondrial calcium uniporter. In addition, it is involved in reducing airway inflammation arises by pepper spray.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
11103-72-3 | |
790.374 | |
JQJSTVUROJELSR-UHFFFAOYSA-H | |
16218584 | |
N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] |
Cl6H46N14O2Ru3 | |
MFCD00011479 | |
6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride | |
azane;ruthenium(2+);hexachloride;dihydrate |
Spécification
>500°C (decomposition) | |
Red | |
Cl6H46N14O2Ru3 | |
MFCD00011479 | |
6cl.o2ru3.14nh3, ruthenium red, technical grade, degrees +/->> aeno>> ie, azane;ruthenium 2+ ;hexachloride;dihydrate, ruthenium red, for microscopy calc. on dry substance, at, ruthenium red, for microscopy calc. on dry basis, at, triruthenoxane-1,1,3,3,5,5-hexakis ylium tetradecaamine hexachloride | |
JQJSTVUROJELSR-UHFFFAOYSA-H | |
azane;ruthenium(2+);hexachloride;dihydrate | |
16218584 | |
≥34.8% Ruthenium | |
3.110 g/mL |
11103-72-3 | |
Crystalline | |
[(NH3)5RuORu(NH3)4ORu(NH3)5]Cl6 | |
14,8300 | |
Soluble in water and ammonium hydroxide. | |
N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+2].[Ru+2].[Ru+2] | |
790.374 | |
786.35 | |
Odorless | |
Ruthenium(III) chloride oxide, Ammoniated |
Sécurité et traitement
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'rtecsNumber' : VM2656500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only