missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylchloride hydrate, Thermo Scientific Chemicals

Product Code. 11314748
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as catalyst. As a precursor to other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32535-76-5
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.42
MDL Number MFCD00049765
InChI Key XTHALOZYNGMAKY-UHFFFAOYSA-K
IUPAC Name trichloro(nitroso)rutheniumtris(ylium)
SMILES Cl[Ru+3](Cl)(Cl)N=O

Specifications

Melting Point 180°C (decomposition)
Physical Form Crystalline
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 237.44 (Anhydrous)
Chemical Name or Material Ruthenium (III) nitrosylchloride hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'