missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(III) nitrosylchloride, Ru 42.1% min, Thermo Scientific Chemicals

Product Code. 11317198
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ruthenium nitrosyl chloride is an important ruthenium compounds, which is used in synthesis of more complex ruthenium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18902-42-6
Molecular Formula Cl3NORu
Molecular Weight (g/mol) 237.43
MDL Number MFCD00049765
InChI Key AUWSZCCJRSCQEP-UHFFFAOYSA-K

Specifications

Physical Form Crystalline
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 237.44
Concentration or Composition (by Analyte or Components) Ru 42.1% min.
Sensitivity Hygroscopic
Chemical Name or Material Ruthenium(III) nitrosylchloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'