Learn More
Ruthenium(III) nitrosylnitrate, Ru 31.3% min, Thermo Scientific Chemicals
Description
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | (decomposition) |
Color | Red to Brown |
Physical Form | Crystalline |
Quantity | 1 g |
Assay Percent Range | Ru 31.3% min. |
Solubility Information | Miscible with cold water. |
Formula Weight | 317.09 |
Odor | Odorless |
Appearance | Red-brown crystals |
Chemical Name or Material | Ruthenium(III) nitrosylnitrate |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.