missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, 99.5+%, anhydrous, Thermo Scientific Chemicals

Product Code. 10647352
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O

Specifications

Color Black
Physical Form Fine Powder
Quantity 5 g
Assay Percent Range 99.5+%
Linear Formula RuO2
Fieser 12,428; 14,272; 15,281
Solubility Information Solubility in water: insoluble
Formula Weight 133.07
Percent Purity ≥99.5%
Packaging Glass bottle
Specific Gravity 6.97
Density 6.9700g/mL
Chemical Name or Material Ruthenium(IV) oxide, 99.5+%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'