Learn More
Ruthenium(IV) oxide, anhydrous, 99.9%, Thermo Scientific Chemicals
Description
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.
Solubility
Insoluble in water, and acids. Soluble in fused alkali.
Notes
Incompatible with oxidizers, acids, aqua regia, organic solvents.

Specifications
Specifications
Melting Point | 1200°C (sublimation) |
Quantity | 5 g |
UN Number | UN1479 |
Solubility Information | Insoluble in water,and acids. Soluble in fused alkali. |
Formula Weight | 133.07 |
Percent Purity | 99.9% |
Odor | Odorless |
Sensitivity | Hygroscopic |
Density | 6.97 |
Assay | (metals basis) |
Show More |
Safety and Handling

- Danger
- Oxidising solid Category 2
- H272-May intensify fire; oxidiser.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.