missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Ru 75.2% min, Thermo Scientific Chemicals

49.55 € - 896.46 €

Chemical Identifiers

CAS 12036-10-1
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00011210
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
Synonym ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
PubChem CID 82848
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11381838
SDS View Documents
Thermo Scientific Alfa Aesar
011804.02
0.5 g
49.55 €
0.50g
Please to purchase this item. Need a web account? Register with us today!
11391838
SDS View Documents
Thermo Scientific Alfa Aesar
011804.04
2 g
179.90 €
2g
Please to purchase this item. Need a web account? Register with us today!
11301848
SDS View Documents
Thermo Scientific Alfa Aesar
011804.09
10 g
896.46 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12036-10-1
133.07
WOCIAKWEIIZHES-UHFFFAOYSA-N
82848
O=[Ru]=O
O2Ru
MFCD00011210
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
dioxoruthenium

Specifications

1200° (sublimation)
Black
99.95% (metals basis)
MFCD00011210
Insoluble in water,and acids. Soluble in fused alkali.
O=[Ru]=O
133.07
133.07
Hygroscopic
6.97
12036-10-1
Powder
O2Ru
ruthenium dioxide, ruthenium iv oxide, ruthenium oxide, ruthenium oxide ruo2, rutheniumoxide ruo2, ruthenium iv dioxide, ruo2, acmc-1brve, ruthenium iv oxide anhydrous
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
82848
Odorless
Black powder
Ruthenium(IV) oxide, anhydrous, Ru 75.2% min.
Videos
Safety and Handling

Safety and Handling

P264b-P280i-P305+P351+P338

H319

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II

missing translation for 'einecsNumber' : 234-840-6

missing translation for 'rtecsNumber' : VM2654000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.