missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium(IV) oxide hydrate, Premion™, 99.99% (metals basis), Ru 54-60%, Thermo Scientific Chemicals

89.75 € - 2070.30 €

Chemical Identifiers

CAS 32740-79-7
Molecular Formula O2Ru
Molecular Weight (g/mol) 133.07
MDL Number MFCD00149846
InChI Key WOCIAKWEIIZHES-UHFFFAOYSA-N
IUPAC Name dioxoruthenium
SMILES O=[Ru]=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11335687
View Documents
Thermo Scientific Alfa Aesar
043403.03
1 g
89.75 €
1g
Please to purchase this item. Need a web account? Register with us today!
11315687
View Documents
Thermo Scientific Alfa Aesar
043403.06
5 g
448.23 €
5g
Please to purchase this item. Need a web account? Register with us today!
11325687
View Documents
Thermo Scientific Alfa Aesar
043403.14
25 g
2070.30 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chemical catalyst, important material for producing resistance and capacitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

32740-79-7
133.07
WOCIAKWEIIZHES-UHFFFAOYSA-N
O=[Ru]=O
O2Ru
MFCD00149846
dioxoruthenium

Specifications

1200°C (sublimation)
Black
O2Ru
UN1479
WOCIAKWEIIZHES-UHFFFAOYSA-N
dioxoruthenium
133.07 (Anhydrous)
Premion™
(metals basis)
32740-79-7
Powder
MFCD00149846
It is soluble in fused alkalis. It is insoluble in water.
O=[Ru]=O
133.07
99.99%
Ru 54 to 60%
Ruthenium (IV) oxide hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H271
May cause fire or explosion
strong oxidizer.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P501c

H315-H319

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

missing translation for 'einecsNumber' : 234-840-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Signal Word
  • Danger

Hazard Category
  • Oxidising solid Category 2

Hazard Statement
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.