Learn More
Ruthenium tetroxide, 0.5% solution in water, stabilized
CAS: 20427-56-9 | O4Ru | 165.07 g/mol
Brand: Thermo Scientific Acros 389000100
139.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
Specifications
Ruthenium tetroxide | |
White | |
10 mL | |
RuO4 | |
15, 8435 | |
GJFMDWMEOCWXGJ-UHFFFAOYSA-N | |
tetraoxoruthenium | |
119079 | |
0.5 % (typically) | |
1 |
20427-56-9, 7732-18-5 | |
Crystalline Powder | |
O4Ru | |
MFCD00074857 | |
ruthenium tetroxide, ruthenium tetraoxide, unii-97e960g9rp, ruthenium viii oxide, ruthenium oxide ruo4, sodium perruthenate, ruthenium oxide ruo4 , t-4, ruo4, 51429-86-8 tri-hydrochloride salt | |
O=[Ru](=O)(=O)=O | |
165.07 | |
165.07 | |
Glass Bottle | |
1.0000g/mL |
Safety and Handling
- Ruthenium tetroxide, 0.5% solution in water
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0464004 | Certificate of Analysis | 28/06/2024 | 389000100 |
A0457368 | Certificate of Analysis | 09/11/2023 | 389000100 |
A0456321 | Certificate of Analysis | 21/09/2023 | 389000100 |
A0445113 | Certificate of Analysis | 25/08/2023 | 389000100 |
A0403359 | Certificate of Analysis | 14/12/2022 | 389000100 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only