missing translation for 'onlineSavingsMsg'
Learn More

Rutin Hydrate, 97+%, Thermo Scientific Chemicals

Product Code. 10071480
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207671-50-9
Molecular Formula C27H30O16
Molecular Weight (g/mol) 610.52
MDL Number MFCD01319140
InChI Key IKGXIBQEEMLURG-NVPNHPEKSA-N
Synonym rutin, rutoside, phytomelin, quercetin 3-rutinoside, birutan, eldrin, myrticolorin, venoruton, 3-rutinosyl quercetin, bioflavonoid
PubChem CID 5280805
ChEBI CHEBI:28527
IUPAC Name 2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(OC4=CC(O)=CC(O)=C4C3=O)C3=CC(O)=C(O)C=C3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 195.0°C
Color White
Loss on Drying 5.5 % to 9.0 % (500mg, 130°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass Bottle
Quantity 5 g
Merck Index 15, 8438
Solubility Information Solubility in water: 1g/8l. Other solubilities: soluble in alkaline solutions,soluble in pyridine and formamide,slightly soluble in alcohol and acetone,practically insoluble in chloroform,carbone,disulfide and benzene
Formula Weight 664.58
Percent Purity 97+%
Physical Form Powder
Chemical Name or Material Rutin hydrate, 97.0-102.0%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'