missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 99%, Thermo Scientific Chemicals

Product Code. 11363916
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine is used as a chiral amine derivative used in studies of real-time chiral discrimination of enantiomers as well as in studies of kinetic resolution of chiral amines with ω-transaminase using an enzyme-membrane reactor.

Solubility
Soluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23357-52-0
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00671630
InChI Key JRZGPXSSNPTNMA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1,2,3,4-tetrahydro-1-naphthylamine, s-+-1,2,3,4-tetrahydro-1-naphthylamine, 1s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1-amino-1,2,3,4-tetrahydronaphthalene, s-+-1-aminotetraline, chembl39537, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1s, s-1,2,3,4-tetrahydro-naphthalen-1-ylamine, s-1-amino-1,2,3,4-tetrahydro-naphthalene
PubChem CID 7058074
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES C1CC(C2=CC=CC=C2C1)N

Specifications

Density 1.035
Boiling Point 118°C to 120°C (8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5645
Quantity 25 g
Beilstein 2360279
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 147.22
Percent Purity ≥99%
Assay ee 99%
Chemical Name or Material (S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.