missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methylphenyl)ethylamine, ChiPros 98%, ee 99+%, Thermo Scientific Chemicals

Product Code. 11329105
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27298-98-2
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00145246
InChI Key UZDDXUMOXKDXNE-UHFFFAOYNA-N
Synonym s-1-p-tolyl ethanamine, s---1-p-tolyl ethylamine, s---4-1-aminoethyl toluene, s-1-p-tolyl ethylamine, 1s-1-4-methylphenyl ethan-1-amine, 1s-1-p-tolyl ethanamine, 1s-1-4-methylphenyl ethanamine, s-1-p-tolylethanamine, 1-4-methylphenyl ethanamine #, s-1-4-methylphenyl ethylamine
PubChem CID 7015759
SMILES CC(N)C1=CC=C(C)C=C1

Specifications

Melting Point <-20°C
Density 0.94
Boiling Point 204°C
Flash Point 82°C (179°F)
Refractive Index 1.5215
Quantity 25 g
UN Number UN2735
Beilstein 2422644
Sensitivity Air Sensitive
IUPAC Name (1S)-1-(4-methylphenyl)ethanamine
Formula Weight 135.21
Percent Purity 98%
Assay ee ≥99%
Chemical Name or Material (S)-(-)-1-(4-Methylphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.