missing translation for 'onlineSavingsMsg'
Learn More

(S)-1-Phenyl-1,3-propanediol, 98%, Thermo Scientific Chemicals

Product Code. 11964651
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96854-34-1
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00221510
InChI Key RRVFYOSEKOTFOG-VIFPVBQESA-N
Synonym 1s-1-phenylpropane-1,3-diol, s-1-phenyl-1,3-propanediol, s-1-phenylpropane-1,3-diol, 1s-phenyl-1,3-propanediol, s-1-phenyl-propane-1,3-diol, s, ?-1-phenyl-1,3-propanediol, 1,3-propanediol,1-phenyl-, 1s, 1,3-propanediol, 1-phenyl-, 1s, s---1-phenyl-1,3-propanediol, puriss sum of enantiomers, gc
PubChem CID 6950537
IUPAC Name (1S)-1-phenylpropane-1,3-diol
SMILES C1=CC=C(C=C1)C(CCO)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 62.0°C to 66.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition −54.50° (22°C c=1,1,2-dichloroethane)
Specific Rotation − 54.50
Formula Weight 152.19
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (S)-1-Phenyl-1, 3-propanediol
Show More Show Less
Product Identifier
  • (S)-1-Phenyl-1,3-propanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.