missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-Phenylethanol, 97+%, Thermo Scientific Chemicals

31.16 € - 426.12 €

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
PubChem CID 443135
ChEBI CHEBI:16346
SMILES C[C@H](O)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329709
View Documents
Thermo Scientific Alfa Aesar
B21188.03
1 g
31.16 €
1g
Please to purchase this item. Need a web account? Register with us today!
11339709
View Documents
Thermo Scientific Alfa Aesar
B21188.06
5 g
105.53 €
5g
Please to purchase this item. Need a web account? Register with us today!
15493227
View Documents
Thermo Scientific Alfa Aesar
B21188.14
25 g
426.12 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of optically active products. Used for the synthesis of various pharmaceutical compounds. It can also be used as a chiral derivatizing agent for determining the absolute configuration of secondary alcohols.

Solubility
Soluble in water at 20°C 20g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

1445-91-6
122.17
WAPNOHKVXSQRPX-ZETCQYMHSA-N
CHEBI:16346
C8H10O
MFCD00064264
443135
C[C@H](O)C1=CC=CC=C1

Specifications

1445-91-6
1.012
85°C (185°F)
C8H10O
MFCD00064264
2039797
Soluble in water at 20°C 20g/L. Soluble in alcohol.
−45° (c=5 in Methanol)
(1S)-1-phenylethanol
443135
122.17
(S)-(-)-1-Phenylethanol
9°C to 11°C
97°C to 99°C (20 mmHg)
Mild
1.528
UN2937
s-1-phenylethanol, s---1-phenylethanol, 1s-1-phenylethanol, 1s-1-phenylethan-1-ol, unii-2mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
WAPNOHKVXSQRPX-ZETCQYMHSA-N
C[C@H](O)C1=CC=CC=C1
122.17
CHEBI:16346
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H227-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ALPHA-METHYLBENZYL ALCOHOL, LIQUID

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.