Learn More
(S)-(-)-1-Phenylethylamine, 98%, Thermo Scientific Chemicals
41.10 € - 353.76 €
Chemical Identifiers
CAS | 2627-86-3 |
---|---|
Molecular Formula | C8H11N |
Molecular Weight (g/mol) | 121.183 |
MDL Number | MFCD00064406 |
InChI Key | RQEUFEKYXDPUSK-ZETCQYMHSA-N |
Synonym | s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine |
PubChem CID | 75818 |
ChEBI | CHEBI:35321 |
IUPAC Name | (1S)-1-phenylethanamine |
SMILES | CC(C1=CC=CC=C1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490463
|
Thermo Scientific Alfa Aesar
A12632.14 |
25 g |
41.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400473
|
Thermo Scientific Alfa Aesar
A12632.22 |
100 g |
113.57 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11410473
|
Thermo Scientific Alfa Aesar
A12632.36 |
500 g |
353.76 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-1-Phenylethylamine is utilized in a one-pot, multi-component synthesis of a highly substituted, chiral pyrrole.
Solubility
Slightly miscible with water.
Notes
Keep away from sources of ignition. Incompatible with acids, acid chlorides, acid anhydrides and strong oxidizing agents.
Chemical Identifiers
2627-86-3 | |
121.183 | |
RQEUFEKYXDPUSK-ZETCQYMHSA-N | |
75818 | |
(1S)-1-phenylethanamine |
C8H11N | |
MFCD00064406 | |
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine | |
CHEBI:35321 | |
CC(C1=CC=CC=C1)N |
Specifications
2627-86-3 | |
0.948 | |
71°C (159°F) | |
C8H11N | |
MFCD00064406 | |
2204907 | |
14,6026 | |
Slightly miscible with water. | |
−30° (c=10 in Ethanol) | |
(1S)-1-phenylethanamine | |
75818 | |
121.18 | |
(S)-(-)-1-Phenylethylamine |
-10°C | |
187°C | |
Amine-like | |
1.526 | |
UN2735 | |
Air Sensitive | |
s---1-phenylethylamine, s-1-phenylethanamine, 1s-1-phenylethanamine, s---alpha-methylbenzylamine, s-1-phenylethylamine, l--alpha-methylbenzylamine, l-alpha-methylbenzylamine, s-alpha-methylbenzenemethanamine, --alpha-phenethylamine, l---1-phenylethylamine | |
RQEUFEKYXDPUSK-ZETCQYMHSA-N | |
CC(C1=CC=CC=C1)N | |
121.183 | |
CHEBI:35321 | |
98% |
Safety and Handling
GHS H Statement
H314-H318-H302-H312-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312-H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 220-098-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- (S)-(-)-1-Phenylethylamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : L-.alpha.-Methylbenzylamin
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.