missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(1-Hydroxyethyl)pyridine, Thermo Scientific Chemicals

Product Code. 15433659
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

 

Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted 2-Pyridinemethanol derivatives are the potent and selective phosphodiesterase-4 inhibitors. Reaction of hydrous nitrosylruthenium trichloride with 2-pyridinemethanol gave ruthenium complexes.

Solubility
TRUSTED_SUSTAINABILITY

Specifications

CAS 59042-90-9
Color Colorless
Molecular Formula C7H9NO
MDL Number MFCD06795465
Quantity 1 g
Synonym s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, 2-s-1-hydroxyethyl pyridine, s---2-1-hydroxyethyl pyridine, s-2-1-hydroxyethyl pyridine, 1s-1-pyridin-2-yl ethan-1-ol, 1s-1-pyridin-2-yl ethanol, s-1-2-pyridyl ethanol, s-2-1-hydroxyethyl pyridine, pubchem5701, 1s-1-pyridin-2-ylethanol
Solubility Information Soluble in toluene.
InChI Key PPHIIIRFJKDTLG-UHFFFAOYNA-N
SMILES CC(O)C1=CC=CC=N1
IUPAC Name (1S)-1-pyridin-2-ylethanol
Molecular Weight (g/mol) 123.16
PubChem CID 11094597
Formula Weight 123.16
Physical Form Solid
Chemical Name or Material (S)-2-(1-Hydroxyethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.