Learn More
(S)-(+)-2-Aminobutane, 98%, Thermo Scientific Chemicals
58.30 € - 709.00 €
Chemical Identifiers
CAS | 513-49-5 |
---|---|
Molecular Formula | C4H11N |
Molecular Weight (g/mol) | 73.139 |
MDL Number | MFCD00064417 |
InChI Key | BHRZNVHARXXAHW-BYPYZUCNSA-N |
Synonym | s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s |
PubChem CID | 6713753 |
IUPAC Name | (2S)-butan-2-amine |
SMILES | CCC(C)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11392417
|
Thermo Scientific Alfa Aesar
L10069.MD |
250 mg |
58.30 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11372417
|
Thermo Scientific Alfa Aesar
L10069.03 |
1 g |
164.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11382417
|
Thermo Scientific Alfa Aesar
L10069.06 |
5 g |
709.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an active pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an active pharmaceutical intermediate.
Solubility
Fully miscible in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
513-49-5 | |
73.139 | |
BHRZNVHARXXAHW-BYPYZUCNSA-N | |
6713753 | |
CCC(C)N |
C4H11N | |
MFCD00064417 | |
s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s | |
(2S)-butan-2-amine |
Specifications
513-49-5 | |
12.5 | |
−19°C (−2°F) | |
C4H11N | |
MFCD00064417 | |
1718760 | |
14,1544 | |
Fully miscible in water. | |
+7.5° (Neat) | |
(2S)-butan-2-amine | |
6713753 | |
98% |
0.722 | |
63°C | |
Amine-like | |
1.393 | |
UN2733 | |
Air Sensitive | |
s-+-2-aminobutane, s-+-sec-butylamine, 2s-butan-2-amine, 2-butanamine, 2s, s-2-butylamine, s-sec-butylamine, +-2-butylamine, unii-z192xwh21o, 2-butanamine, s | |
BHRZNVHARXXAHW-BYPYZUCNSA-N | |
CCC(C)N | |
73.139 | |
73.14 | |
(S)-(+)-2-Aminobutane |
Safety and Handling
GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 208-164-7
missing translation for 'rtecsNumber' : EO3327000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only