missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Aminoethyl)isothiourea dihydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11369065
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-10-0
Molecular Formula C3H11Br2N3S
Molecular Weight (g/mol) 281.01
MDL Number MFCD00037011
InChI Key XDVMCVGTDUKDHL-UHFFFAOYSA-N
Synonym 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide
PubChem CID 5940
IUPAC Name 2-aminoethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N.Br.Br

Specifications

Melting Point 190°C to 192°C
Odor Odorless
Quantity 5 g
Beilstein 3911163
Merck Index 14,178
Formula Weight 281.02
Percent Purity 98%
Chemical Name or Material S-(2-Aminoethyl)isothiourea dihydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'