missing translation for 'onlineSavingsMsg'
Learn More
S-(2-Aminoethyl)isothiourea dihydrobromide, 98%, Thermo Scientific Chemicals Product Code.: 11379065

S-(2-Aminoethyl)isothiourea dihydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11379065
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11379065

Brand: Thermo Scientific Alfa Aesar L14232.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-10-0
Molecular Formula C3H11Br2N3S
Molecular Weight (g/mol) 281.01
MDL Number MFCD00037011
InChI Key XDVMCVGTDUKDHL-UHFFFAOYSA-N
Synonym 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide
PubChem CID 5940
IUPAC Name 2-aminoethyl carbamimidothioate;dihydrobromide
SMILES C(CSC(=N)N)N.Br.Br

Specifications

Melting Point 190°C to 192°C
Odor Odorless
Quantity 25 g
Beilstein 3911163
Merck Index 14,178
Formula Weight 281.02
Percent Purity 98%
Chemical Name or Material S-(2-Aminoethyl)isothiourea dihydrobromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM22K003 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog NumberL14232.06,L14232.14
Lot NumberU09J007 Certificate TypeCertificate of Analysis Date21/08/2023 Catalog NumberL14232.06,L14232.14
Lot NumberS18H018 Certificate TypeCertificate of Analysis Date30/11/2022 Catalog NumberL14232.06,L14232.14
Lot NumberP24H016 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberL14232.06,L14232.14
Lot NumberL01U035 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14232.06,L14232.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S-(2-Aminoethyl)isothiourea dihydrobromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.