missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Aminomethyl)pyrrolidine, 98%, Thermo Scientific Chemicals

Product Code. 10537064
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69500-64-7
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.17
MDL Number MFCD00191745
InChI Key AUKXFNABVHIUAC-UHFFFAOYNA-N
Synonym s-pyrrolidin-2-ylmethanamine, s-+-2-aminomethyl pyrrolidine, s-2-aminomethyl pyrrolidine, 2s-pyrrolidin-2-ylmethylamine, s-pyrrolidin-2-ylmethylamine, s-2-aminomethylpyrrolidine, 2s-pyrrolidin-2-yl methanamine, 2-pyrrolidinemethanamine, 2s, 1-2s-pyrrolidin-2-yl methanamine, d07vdj
PubChem CID 2734054
ChEBI CHEBI:44632
IUPAC Name [(2S)-pyrrolidin-2-yl]methanamine
SMILES NCC1CCCN1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.933g/mL
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 mg
Formula Weight 100.16
Chemical Name or Material (S)-(+)-2-(Aminomethyl)pyrrolidine
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-2-(Aminomethyl)pyrrolidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.