missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminononane, ChiPros 99+%, ee 99+%, Thermo Scientific Chemicals

Product Code. 11334226
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869278-88-6
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD03844738
InChI Key ALXIFCUEJWCQQL-UHFFFAOYNA-N
Synonym 2-nonanamine, 2-aminononane, 2-nonylamine, 2-amino-nonane, acmc-20apfg, acmc-1bh6g
PubChem CID 12878166
IUPAC Name nonan-2-amine
SMILES CCCCCCCC(C)N

Specifications

Density 0.782
Boiling Point 73°C (19 mmHg)
Flash Point 71°C (159°F)
Refractive Index 1.4271
Quantity 5 g
UN Number UN2735
Sensitivity Air Sensitive
Optical Rotation +6° (Neat)
Formula Weight 143.27
Percent Purity ≥99%
Assay ee ≥99%
Chemical Name or Material (S)-(+)-2-Aminononane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.