Learn More
(S)-(-)-2-Methylbutylamine, 98+%, Thermo Scientific Chemicals
54.67 € - 177.89 €
Chemical Identifiers
CAS | 34985-37-0 |
---|---|
Molecular Formula | C5H13N |
Molecular Weight (g/mol) | 87.166 |
MDL Number | MFCD00064430 |
InChI Key | VJROPLWGFCORRM-YFKPBYRVSA-N |
Synonym | s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride |
PubChem CID | 2724272 |
IUPAC Name | (2S)-2-methylbutan-1-amine |
SMILES | CCC(C)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318945
|
Thermo Scientific Alfa Aesar
L14063.MD |
250 mg |
54.67 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308945
|
Thermo Scientific Alfa Aesar
L14063.03 |
1 g |
177.89 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(-)-2-Methylbutylamine is used as aintermediate in organic synthesis, as pharmaceutical intermediate.
Solubility
Fully miscible in water.
Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air, carbondioxide.
Chemical Identifiers
34985-37-0 | |
87.166 | |
VJROPLWGFCORRM-YFKPBYRVSA-N | |
2724272 | |
CCC(C)CN |
C5H13N | |
MFCD00064430 | |
s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride | |
(2S)-2-methylbutan-1-amine |
Specifications
34985-37-0 | |
3°C (37°F) | |
MFCD00064430 | |
1718807 | |
VJROPLWGFCORRM-YFKPBYRVSA-N | |
CCC(C)CN | |
87.166 | |
87.16 | |
(S)-(-)-2-Methylbutylamine |
96°C to 97°C | |
C5H13N | |
UN2733 | |
s---2-methylbutylamine, s-2-methylbutylamine, unii-d8q8rzf45x, 2s-2-methylbutan-1-amine, s---2-methyl-1-butylamine, d8q8rzf45x, 2s-2-methyl-butan-1-amine, 2-methyl-1-butanamine #, s---1-amino-2-methylbutane hydrochloride | |
−5.9° (Neat) | |
(2S)-2-methylbutan-1-amine | |
2724272 | |
≥98% |
Safety and Handling
GHS H Statement
H225-H314-H318-H335
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
- (S)-(-)-2-Methylbutylamine
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only