missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11392577

(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11392577
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11392577

Brand: Thermo Scientific Alfa Aesar L11711.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid is used as a building block for pharmaceuticals and asymmetric synthesis. It Is also used as a resolving agent for chiral bases and in an efficient synthesis of quinolizidine and indolizidine alkaloids.

Solubility
Soluble in water (10-15% at 20°C), alcohol, acetone, dimethyl sufoxide and methanol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxoShow MoreShow Less
PubChem CID 7405
ChEBI CHEBI:18183
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 158°C to 162°C
Quantity 500 g
Beilstein 82132
Merck Index 14,8001
Solubility Information Soluble in water (10-15% at 20°C),alcohol,acetone,dimethyl sufoxide and methanol.
Optical Rotation −10.5° (c=5 in Water)
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
Formula Weight 129.12
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Pyrrolidinone-5-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162574 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog NumberL11711.14,L11711.36,L11711.22
Lot Number10164130 Certificate TypeCertificate of Analysis Date23/11/2023 Catalog NumberL11711.14,L11711.36,L11711.22
Lot Number10164130 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL11711.14,L11711.36,L11711.22
Lot NumberUSLY000034 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL11711.14,L11711.36,L11711.22
Lot Number10113455 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL11711.14,L11711.36,L11711.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.