missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 10148053

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10148053
25 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10148053

Brand: Thermo Scientific Acros 132120250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-79-3
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00005272
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym l-pyroglutamic acid, pyroglutamic acid, pidolic acid, 5-oxo-l-proline, h-pyr-oh, 5-oxoproline, s-5-oxopyrrolidine-2-carboxylic acid, 2s-5-oxopyrrolidine-2-carboxylic acid, l-5-oxoproline, l-proline, 5-oxoShow MoreShow Less
PubChem CID 7405
ChEBI CHEBI:18183
IUPAC Name (2S)-5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Melting Point 152.0°C to 162.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 25 g
Beilstein 22,284
Merck Index 15,8113
Specific Rotation Condition − 27.50 (20.00°C c=10,1 N NaOH)
Specific Rotation − 27.50
Solubility Information Solubility in water: 100-150g/l (20°). Other solubilities: soluble in alcohol and acetone
Formula Weight 129.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Pyrrolidone-5-carboxylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0400192 Certificate TypeCertificate of Analysis Date04/09/2023 Catalog Number132121000,132120250,132125000
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-2-Pyrrolidone-5-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.