missing translation for 'onlineSavingsMsg'
Learn More

(S)-3,5-DHPG, Tocris Bioscience™

Product Code. 18745936
Click to view available options
Quantity:
5 mg
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective group I mGlu agonist; active enantiomer of 3,5-DHPG (Cat. No. 0342)

(S)-3,5-DHPG is a selective group I mGlu receptor agonist. (S)-3,5-DHPG regulates the expression of microRNAs in mouse cerebral cortex in vivo. (S)-3,5-DHPG induces a profound depression of synaptic dopamine release in mouse cortico-striatal cells in vitro, and anxiety-like behavior in vivo in mice.

Racemate also available.

Chemical Identifiers

CAS 162870-29-3
Molecular Formula C8H9NO4
Molecular Weight (g/mol) 183.16
MDL Number MFCD11044457
InChI Key HOOWCUZPEFNHDT-UHFFFAOYNA-N
Synonym s-3,5-dihydroxyphenylglycine, s-3,5-dhpg, l-3,5-dihydroxyphenylglycine, s-dhpg, unii-cf5g2g268a, chembl39221, 2s-2-amino-2-3,5-dihydroxyphenyl acetic acid, benzeneacetic acid, alpha-amino-3,5-dihydroxy-, alphas, tocris-0342, tocris-0805
PubChem CID 443586
ChEBI CHEBI:29474
IUPAC Name 2-amino-2-(3,5-dihydroxyphenyl)acetic acid
SMILES NC(C(O)=O)C1=CC(O)=CC(O)=C1

Specifications

Formula Weight 183.16
Percent Purity >98%
Quantity 5 mg
Chemical Name or Material (S)-3, 5-DHPG
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'