missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ (S)-(+)-3-Aminopyrrolidine dihydrochloride, 98%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Melting Point | ∽283°C (decomposition) |
Quantity | 250 mg |
Beilstein | 4870016 |
Sensitivity | Hygroscopic |
Optical Rotation | +1.2° (c=10 in Water) |
Formula Weight | 159.06 |
Percent Purity | 98% |
Chemical Name or Material | (S)-(+)-3-Aminopyrrolidine dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'