missing translation for 'onlineSavingsMsg'
Learn More
(S)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific™ Product Code.: 15585936

(S)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific™

Product Code. 15585936
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15585936

Brand: Thermo Scientific Acros 456930050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 204512-95-8
Molecular Formula C4H9NO·ClH
Molecular Weight (g/mol) 123.58
InChI Key MHOVLDXJDIEEMJ-WCCKRBBISA-N
Synonym s-tetrahydrofuran-3-amine hydrochloride, s-3-aminotetrahydrofuran hydrochloride, 3s-oxolan-3-amine hydrochloride, s-3-aminotetrahydrofuran hcl, s-tetrahydrofuran-3-amine hcl, s-tetrahydro-furan-3-ylamine hydrochloride, 3-furanamine, tetrahydro-, hydrochloride, 3s, s-3-aminotetrahydrofuranhydrochloride, pubchem15126, ksc916a5tShow MoreShow Less
PubChem CID 18664284
IUPAC Name (3S)-oxolan-3-amine;hydrochloride
SMILES C1COCC1N.Cl

Specifications

Melting Point 140.0°C to 145.0°C
Infrared Spectrum Authentic
Assay Percent Range 94.0 to max. 106% min. (Argentometry)
Quantity 5 g
Specific Rotation -7.4
Formula Weight 123.58
Percent Purity 95%
Chemical Name or Material (S)-3-Aminotetrahydrofuran hydrochloride
compliance-icons
Product Identifier
  • (S)-3-Aminotetrahydrofuran hydrochloride, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.