missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Butyn-2-ol, 95%, 98% ee, Thermo Scientific Chemicals

Product Code. 10643573
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2914-69-4
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00190166
InChI Key GKPOMITUDGXOSB-BYPYZUCNSA-N
Synonym s---3-butyn-2-ol, s-3-butyn-2-ol, 2s-but-3-yn-2-ol, s-but-3-yn-2-ol, 3-butyn-2-ol, 2s, s---1-butyn-3-ol, 2s-3-butyn-2-ol, 3r-+-3-hydroxybut-1-yne, s---3-butyn-2-ol, purum gc
PubChem CID 6995470
IUPAC Name (2S)-but-3-yn-2-ol
SMILES CC(C#C)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8940g/mL
Boiling Point 108°C to 111°C
Flash Point 26°C
Enantiomeric Excess 97.5% min. (GC)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 0.894
Formula Weight 70.09
Percent Purity 95%,98% ee
Physical Form Liquid
Chemical Name or Material (S)-(-)-3-Butyn-2-ol, 95%, (98% e.e.)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(-)-3-Butyn-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'