missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11379575
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36567-72-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00145262
InChI Key AYOLELPCNDVZKZ-QMMMGPOBSA-N
Synonym s-3-hydroxy-3-phenylpropanoic acid, 3s-3-hydroxy-3-phenylpropanoic acid, s-3-hydroxy-3-phenylpropionic acid, ?-3-phenylhydracrylic acid, s---3-hydroxy-3-phenylpropionic acid, benzenepropanoic acid,, a-hydroxy-,, as, s---3-hydroxy-3-phenylpropionic acid, 98+%, unii-s2w4lws09c component, s-3-hydroxy-3-phenylpropionic acid sum of enantiomers, gc
PubChem CID 2735057
ChEBI CHEBI:51058
IUPAC Name (3S)-3-hydroxy-3-phenylpropanoic acid
SMILES C1=CC=C(C=C1)C(CC(=O)O)O

Specifications

Melting Point 117°C to 119°C
Quantity 250 mg
Beilstein 3197773
Optical Rotation −22° (c=4 in Methanol)
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-3-Hydroxy-3-phenylpropionic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'