missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Safety Waste Caps
GREENER_CHOICE

Safety Waste Caps for the safe disposal of liquid waste. For maximum protection and efficient work with hazardous waste liquids. Offers multiple options to connect different tubing and capillary sizes.

Brand:  SCAT™ 107051

113.36 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10880191

  • 144.00 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 360° free turning PTFE body, resistant against aggressive media
  • Exhaust filter system against harmful vapors
  • Fittings and tube connectors in different sizes
  • Flexible system
  • Time saving in changing the waste containers while tubes are fixed
  • Lab safety, technical protective measures, optimal protection when collecting liquid waste
  • Cost efficiency
  • Huge range of accessories, optional: funnel use, level control preventing overflowing containers
  • Huge portfolio of thread sizes suitable for waste containers from different manufacturers
  • Safety disposal of HPLC waste
  • High work performance
  • Personal protection, avoid skin contact with organic solvents, occupational safety, ergonomics
  • Sealed bottles and containers
  • Create a comfortable working climate
Specifications

Specifications

White
Waste Disposal Container
PTFE
Safety Waste Cap
HPLC Disposal Systems
3 x 2.3/3.2 mm OD, Connector for Exhaust Filter
B63
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.