missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(-)-Carbidopa, Tocris Bioscience™
Click to view available options
Quantity:
25 mg
Description
Peripheral decarboxylase inhibitor, used in combination with levodopa for treatment of Parkinsonism.
Specifications
Specifications
CAS | 28860-95-9 |
Molecular Formula | C10H14N2O4 |
Quantity | 25 mg |
Synonym | carbidopa, lodosyn, n-aminomethyldopa, carbidopum, s---carbidopa, s-carbidopa, alpha-methyldopahydrazine, l-alpha-methyldopahydrazine, carbidopa anhydrous |
InChI Key | TZFNLOMSOLWIDK-JTQLQIEISA-N |
SMILES | CC(CC1=CC(=C(C=C1)O)O)(C(=O)O)NN |
IUPAC Name | (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid |
Molecular Weight (g/mol) | 226.232 |
PubChem CID | 34359 |
ChEBI | CHEBI:39585 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction