Learn More
(S)-(+)-Mandelic Acid ≥99%, Thermo Scientific Chemicals
28.84 € - 58.59 €
Chemical Identifiers
CAS | 17199-29-0 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00004495 |
InChI Key | IWYDHOAUDWTVEP-ZETCQYMHSA-N |
Synonym | s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid |
PubChem CID | 439616 |
ChEBI | CHEBI:32800 |
IUPAC Name | (2S)-2-hydroxy-2-phenylacetic acid |
SMILES | C1=CC=C(C=C1)C(C(=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10386810
|
Thermo Scientific Acros
125300250 |
25 g | Plastic bottle |
28.84 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396810
|
Thermo Scientific Acros
125301000 |
100 g | Plastic bottle |
58.59 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
17199-29-0 | |
152.149 | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
439616 | |
(2S)-2-hydroxy-2-phenylacetic acid |
C8H8O3 | |
MFCD00004495 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
CHEBI:32800 | |
C1=CC=C(C=C1)C(C(=O)O)O |
Specifications
17199-29-0 | |
99.0 | |
Beige-Brown to White | |
C8H8O3 | |
MFCD00004495 | |
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid | |
C1=CC=C(C=C1)C(C(=O)O)O | |
152.149 | |
CHEBI:32800 | |
≥99.0% (Acid) | |
Crystalline Powder |
130°C to 135°C | |
100.0 | |
99+% | |
C6H5CH(OH)CO2H | |
10, 192 | |
IWYDHOAUDWTVEP-ZETCQYMHSA-N | |
(2S)-2-hydroxy-2-phenylacetic acid | |
439616 | |
152.15 | |
Authentic | |
(S)-(+)-Mandelic acid, 99+% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 241-240-8
missing translation for 'tsca' : TSCA
- Lead(II) acetate trihydrate
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 1A
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H360Df-May damage the unborn child. Suspected of damaging fertility.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only