missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Mandelic acid, 99+%, Thermo Scientific Chemicals

27.03 € - 180.90 €

Chemical Identifiers

CAS 17199-29-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00004495
InChI Key IWYDHOAUDWTVEP-ZETCQYMHSA-N
Synonym s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
PubChem CID 439616
ChEBI CHEBI:32800
IUPAC Name (2S)-2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11487337
View Documents
Thermo Scientific Alfa Aesar
A15061.06
5 g
27.03 €
5g
Please to purchase this item. Need a web account? Register with us today!
11497337
View Documents
Thermo Scientific Alfa Aesar
A15061.14
25 g
65.73 €
25g
Please to purchase this item. Need a web account? Register with us today!
11407347
View Documents
Thermo Scientific Alfa Aesar
A15061.22
100 g
180.90 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Mandelic acid is a versatile reagent used in the resolution of racemates and the preparation of amides, Pharmaceutical Intermediates.

Solubility
Soluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

17199-29-0
152.149
IWYDHOAUDWTVEP-ZETCQYMHSA-N
439616
(2S)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00004495
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
CHEBI:32800
C1=CC=C(C=C1)C(C(=O)O)O

Specifications

17199-29-0
C8H8O3
s-+-mandelic acid, s-mandelic acid, s-2-hydroxy-2-phenylacetic acid, l-mandelic acid, l-+-mandelic acid, 2s-2-hydroxy-2-phenylacetic acid, mandelic acid, s, unii-l0umw58g3t, l +-mandelic acid
IWYDHOAUDWTVEP-ZETCQYMHSA-N
(2S)-2-hydroxy-2-phenylacetic acid
439616
152.15
+155° (c=5 in Water)
(S)-(+)-Mandelic acid
131°C to 135°C
MFCD00004495
Soluble in water.
C1=CC=C(C=C1)C(C(=O)O)O
152.149
CHEBI:32800
≥99%
Light sensitive
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

P280-P305+P351+P338-P310

H318

missing translation for 'einecsNumber' : 241-240-8

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only