missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asecos™ S-PHOENIX-90 Light Gray Safety Storage Cabinets with Door Open Arrest System
Safety storage cabinets with folding doors for unrestricted storage of flammable hazardous materials in work areas in accordance with EN 14470-1 and TRGS 510.
Brand: Asecos™ 3011800133715
This item is not returnable.
View return policy
Description
- Optimal developed door technology for easy operation
- Air-dampened door buffer ensures soft and low-wear closings
- Available in a range of widths and heights
- Space-saving folding doors – ideal for use in case of narrow passage widths in front of the cabinet
- Convenient handling – door operation with minimal effort
- Door open arrest system allows convenient and safe removal or storage of containers with both hands
- Continuous anti-trapping door strips on the folding doors offer maximum protection during operation
- Integrated adjusting aids in the cabinet base ensure a fast and effective compensation for uneven floors of up to 10 mm
- Available with different combinations of drawers, height-adjustable shelves and bottom collecting sumps for leakages
- Fire resistance of 90 minutes
- Robust construction and longevity – triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Integrated air ducts ready for connection (DN 75) to a technical exhaust system
- One-hand operation for double-door cabinets
Specifications
EN 14470-1, TRGS 510 | |
616 x 1196 x 1968 mm | |
Storage of flammable hazardous materials | |
2 | |
462.5 kg | |
Steel |
3 x 75 kg sheet steel shelves; 1 x 75 kg sheet steel insert; 1 x 33 L sheet steel sump | |
Folding | |
Shelves: 75 kg each, Insert: 75 kg, Sump: 33 L, Max load: 600 kg | |
Powder-coated | |
Light Gray | |
Safety storage cabinet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction