missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-sec-Phenethyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10712261
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chiral derivatization reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-91-6
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00064264
InChI Key WAPNOHKVXSQRPX-ZETCQYMHSA-N
Synonym s-1-phenylethanol, s---1-phenylethanol, 1s-1-phenylethanol, 1s-1-phenylethan-1-ol, unii-2mic4qly2x, 1-phenylethanol, s, 2mic4qly2x, s-1-phenethyl alcohol, s-1-phenyl-1-ethanol, s-1-phenylethan-1-ol
PubChem CID 443135
ChEBI CHEBI:16346
SMILES C[C@H](O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 9°C to 11°C
Color Colorless
Density 1.0100g/mL
Boiling Point 98°C (20.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)OH
Refractive Index 1.5260 to 1.528
Quantity 1 g
Beilstein 06, IV, 3029
Specific Gravity 1.01
Specific Rotation Condition −42.50 (20.00°C neat)
Specific Rotation -42.5
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in glycerol
IUPAC Name (1S)-1-phenylethanol
Formula Weight 122.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-(-)-sec-Phenethyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-sec-Phenethyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'