missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-Tetrahydro-3-furoic acid, 97%, Thermo Scientific Chemicals Product Code.: 15472739

(S)-(-)-Tetrahydro-3-furoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15472739
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 15472739

Brand: Thermo Scientific Alfa Aesar H66392.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 168395-26-4
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD07369984
InChI Key BOTREHHXSQGWTR-UHFFFAOYNA-N
Synonym s-tetrahydrofuran-3-carboxylic acid, 3s-oxolane-3-carboxylic acid, s-tetrahydro-3-furancarboxylic acid, s-tetrahydro-3-furoic acid, s-tetrahydro-furan-3-carboxylic acid, 3s-tetrahydrofuran-3-carboxylic acid, l-tetrahydrofuran-3-carboxylic acid, 3-furancarboxylic acid, tetrahydro-, 3s, 3s-3-oxolanecarboxylic acid, s, ?-tetrahydro-3-furancarboxylic acidShow MoreShow Less
PubChem CID 40784874
IUPAC Name oxolane-3-carboxylic acid
SMILES OC(=O)C1CCOC1

Specifications

Color Yellow to Brown
Quantity 1 g
Beilstein 1281203
Formula Weight 116.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(-)-Tetrahydro-3-furoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ29A050 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66392.03
Lot NumberQ16H002 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66392.03
Lot NumberT21D031 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberH66392.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-Tetrahydro-3-furoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.