missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(-)-S-Trityl-D-cysteine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.
Solubility
Sparingly soluble in water 0.022 g/L 25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
(-)-S-Trityl-D-cysteine is used in medicine as intermediate. It inhibits the human mitotic kinesin Eg5.
Solubility
Sparingly soluble in water 0.022 g/L 25°C.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.

Specifications
Specifications
Solubility Information | Sparingly soluble in water 0.022g/L 25°C. |
Quantity | 1 g |
Formula Weight | 363.47 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | (-)-S-Trityl-D-cysteine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(-)-S-Trityl-D-cysteine, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction