missing translation for 'onlineSavingsMsg'
Learn More

(S)-verbenone, Thermo Scientific™

Product Code. 10199652
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

Density 0.9700g/mL
Assay Percent Range 97%
Quantity 1 g
Specific Rotation Condition − 220.00 (20.00°C c=3−4,CHCL3)
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material (S)-Verbenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'