missing translation for 'onlineSavingsMsg'
Learn More

(S)-verbenone, Thermo Scientific™

Product Code. 10537953
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1196-01-6
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001343
InChI Key DCSCXTJOXBUFGB-JGVFFNPUSA-N
Synonym levoverbenone, --verbenone, l-verbenone, 1s---verbenone, --2-pinen-4-one, verbenone, unii-2xp0j7754u, 1s,5s-4,6,6-trimethylbicyclo 3.1.1 hept-3-en-2-one, verbenone, l, bicyclo 3.1.1 hept-3-en-2-one, 4,6,6-trimethyl-, 1s,5s
PubChem CID 92874
ChEBI CHEBI:78316
IUPAC Name (1S,5S)-2,6,6-trimethylbicyclo[3.1.1]hept-2-en-4-one
SMILES CC1=CC(=O)[C@H]2C[C@@H]1C2(C)C

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material (S)-Verbenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.