missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A2 293T Cell Overexpression Lysate (Denatured), Abnova
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Western Blotting
Specifications
Specifications
| Cell Line | 293T |
| For Use With (Application) | Western Blot |
| Description | S100A2 293T Cell Transient Overexpression Lysate(Denatured) |
| Gene Accession No. | BC002829 |
| Molecular Weight (g/mol) | 10.78 |
| pH Range | 6.8 |
| Plasmid | pCMV-S100A2 full-length |
| Quality Control Testing | Transient overexpression cell Lysate was tested with Anti-S100A2 antibody (H00006273-B01) by western blotting |
| Quantity | 100 μL |
| Storage Buffer | 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction