missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar

Product Code. p-4522926
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
90mm Plate
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11963662

Brand: Thermo Scientific™ PO0410B

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, on a ready-to-use prepared media plate.

Isolate, cultivate and differentiate dermatophytes, fungi and yeasts, such as Candida albicans, using acidic pH Thermo Scientific™ Sabouraud Glucose Agar.

Fungi maintain their typical cultural appearance and may thus be readily identified according to the standard macroscopic characteristics. Infections caused by Candida spp. are diverse, ranging from superficial and local to deep-seated and disseminated. Candida albicans and Candida glabrata comprise 70-80% of yeast isolates recovered from patients with invasive disease. This culture media, also available in powder form, has applications in environmental monitoring and sterility testing, enabling corrective measures to be implemented rapidly if unacceptable levels of microbial loading indicate lapses in process control. 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Color Straw, clear
Content And Storage 2 to 25°C
Format 90mm Plate
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 19.5 ± 1.0g
Packaging Type Triple Wrapped - Irradiated
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.