missing translation for 'onlineSavingsMsg'
Learn More

Saclofen, Tocris Bioscience™

Product Code. 30001067
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective GABAB antagonist

Selective antagonist at GABAB receptors.

Chemical Identifiers

CAS 125464-42-8
Molecular Formula C9H12ClNO3S
Molecular Weight (g/mol) 249.71
MDL Number MFCD00216817
InChI Key JYLNVJYYQQXNEK-UHFFFAOYNA-N
Synonym saclofen, 3-amino-2-4-chlorophenyl propane-1-sulfonic acid, beta-aminomethyl-4-chlorobenzeneethanesulfonic acid, rs-3-amino-2-4-chlorophenyl propylsulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro, saclofen, solid, saclofen hcl, biomol-nt_000242, d0s5yh, 3-amino-2-4-chlorophenyl-1-propanesulfonic acid
PubChem CID 122150
IUPAC Name 3-amino-2-(4-chlorophenyl)propane-1-sulfonic acid
SMILES NCC(CS(O)(=O)=O)C1=CC=C(Cl)C=C1

Specifications

Quantity 10 mg
Formula Weight 249.72
Percent Purity >99%
Chemical Name or Material Saclofen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.