Learn More
Brady™ SafeKey Padlocks - aluminum with 38 mm Steel Shackle, Keyed Different
The ultimate locks for indoor and outdoor heavy duty safety applications.
33.90 € - 208.00 €
Specifications
Product Type | Keyed Padlocks |
---|---|
Key Type | Keyed Different |
Material | Anodized Aluminum |
Diameter (English) Shackle | 0.25 in. |
Diameter (Metric) Shackle | 6.4 mm |
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
17170291
|
Brady™
150242 |
1 Ea. | Silver |
33.90 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17180291
|
Brady™
150243 |
1 Ea. | Black |
33.90 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
17100311
|
Brady™
150263 |
1 Ea. | Orange |
33.90 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
17110311
|
Brady™
150264 |
1 Ea. | Green |
33.90 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17100331
|
Brady™
150286 |
1 Ea. | Brown |
33.90 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17110331
|
Brady™
150287 |
1 Ea. | Blue |
33.90 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
17140361
|
Brady™
150326 |
6 Pk. | Orange |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17120331
|
Brady™
150288 |
1 Ea. | Yellow |
33.90 €
Each |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17180341
|
Brady™
150307 |
1 Ea. | Red |
33.90 €
Each |
In Stock
Log in to see stock. |
||||
17180361
|
Brady™
150333 |
1 Ea. | Purple |
33.90 €
Each |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
17190271
|
Brady™
150219 |
6 Pk. | Yellow |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17150291
|
Brady™
150240 |
6 Pk. | Black |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17160301
|
Brady™
150256 |
6 Pk. | Green |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17140321
|
Brady™
150278 |
6 Pk. | Silver |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17120341
|
Brady™
150301 |
6 Pk. | Brown |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17130341
|
Brady™
150302 |
6 Pk. | Blue |
200.00 €
Pack of 6 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
||||
17140341
|
Brady™
150303 |
6 Pk. | Red |
208.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
17180371
|
Brady™
150347 |
6 Pk. | Purple |
200.00 €
Pack of 6 |
Estimated Shipment: 12-02-2025
Log in to see stock. |
||||
Description
- Manufactured with solid aluminum body and steel shackle, Brady™'s padlocks are lightweight and rugged
- Compliments the Brady™ SafeKey Padlock range and have the same features
- Locks are chemical and corrosion resistant
- The unique lock mechanism is insulated and has 6 precision steel tumbler blades for a high number of key differences and tamper resistance
- The colour-matched key and lock body allows a fast identification in the application
- 1 key is supplied with each lock
- Custom keying, engraving, charting, Master Key and Grand Master Key are available
Specifications
Keyed Padlocks | |
Anodized Aluminum | |
6.4 mm | |
38 mm | |
6 | |
1.3 in. | |
33 mm |
Keyed Different | |
0.25 in. | |
1.5 in. | |
Steel | |
1.5 in. | |
38 mm |