Learn More
DuPont™ Tyvek 500 Xpert Hooded Coverall
GREENER_CHOICE
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
11.45 € - 1210.00 €
Specifications
Color White
Seam Type Stitched
View More Specs
Products 23
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
13015683
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663953
Small
12.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11743103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663953
Small
954.00 €
Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
12971345
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663953
Small
256.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
13412429
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12991345
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium
256.00 €
Pack of 25
In Stock
to see stock.
Add to basket
11703103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium
954.00 €
Pack of 100
Estimated Shipment: 17-02-2025
to see stock.
Add to basket
13005683
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663967
Medium
11.45 €
Each
In Stock
to see stock.
Edge
Add to basket
11713103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663977
Large
954.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
13095673
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663977
Large
11.45 €
Each
In Stock
to see stock.
Edge
Add to basket
12911355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663977
Large
256.00 €
Pack of 25
In Stock
to see stock.
Add to basket
11723103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663986
X-Large
954.00 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
12931355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663986
X-Large
256.00 €
Pack of 25
In Stock
to see stock.
Add to basket
13025683
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663986
X-Large
11.45 €
Each
In Stock
to see stock.
Edge
Add to basket
12951355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663997
2X-Large
256.00 €
Pack of 25
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
13075673
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663997
2X-Large
11.45 €
Each
In Stock
to see stock.
Edge
Add to basket
11733103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14663997
2X-Large
954.00 €
Pack of 100
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
12961355
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
303.00 €
Pack of 25
In Stock
to see stock.
Add to basket
11753103
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
965.00 €
Pack of 100
In Stock
to see stock.
Add to basket
13085673
GREENER_CHOICE
View accessory products View Documents
DuPont™
D14664003
3X-Large
12.25 €
Each
In Stock
to see stock.
Edge
Add to basket
17402893
View Documents
DuPont™
D15553613
4X-Large
1065.00 €
Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
17412893
View Documents
DuPont™
D15553614
5X-Large
1115.00 €
Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
17422893
View Documents
DuPont™
D15553615
6X-Large
1160.00 €
Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
17432893
View Documents
DuPont™
D15553616
7X-Large
1210.00 €
Pack of 100
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
Description
Description
Composed of flash spun high density polyethylene. Dupont™ Tyvek™ Classic Xpert Hooded Coverall is permeable to both air and water vapor, yet repels water-based liquids and aerosols and offers an excellent barrier against fine particles and fibers.
  • Robust yet lightweight (<180g per garment)
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Ample crotch area for freedom of movement
  • Large, easy-to-grasp zipper puller
  • 3-piece hood for optimal fit to head and face when turning
  • White, sizes S to XXXL
Specifications
Specifications
White
Stitched
Videos
SDS
Documents
Documents
Product Certifications
The user should make the risk assessment to determine the protection required.