missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Safety Bottle Carrier

The Safety Bottle Carrier provides a safe and reliable way to transport a variety of bottled reagents in your lab.

Brand:  Fisherbrand™ 120916F

 View more versions of this product

Product Code. 17800782

  • 67.75 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Robust Design
  • Ergonomic handle prevents hand fatigue and related accidents when carrying heavier loads
  • Handle can be pressed down to either side for easier loading and unloading
  • Centering ring stabilizes bottle in the center of carrier for safer, steadier transport
  • Carriers come in four colors with designated labeling areas for easy identification
  • Made of chemical resistant thermoplastic elastomer - PP blend
Specifications

Specifications

1 Safety Bottle Carrier vessel and attachable handle
Safety Bottle Carrier, 1.5 L, Black
Not Autoclavable
Bottles
1
None
Non-sterile
1.5 L bottles
-20 to 60°C
11.4 cm
11.4 cm
.25 kg
Bottle Holder
1 x 1
Black
Polypropylene
Cylindrical
No
Transportation of bottled regeants
1.5 L
11.4 cm
16.5 cm (25.4 cm Height, Handle Extended)
11.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers